Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syp
Entrez Gene ID
42460
Gene Name
Syncrip
Gene Aliases
AI945337, CG17838, Dmel\CG17838, Dmel_CG17838, anon-WO0118547.613, cg17838, l(3)03806, syp
Chromosome Location
Chr.3R: 20756729 - 20810726 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0038826
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275847.1 NP_001262776.1 2 - 3 607 68
NM_001275849.1 NP_001262778.1 2 - 3 607 68
NM_001275850.1 NP_001262779.1 2 - 3 607 68
NM_001369986.1 NP_001356920.1 2 - 3 607 68
NM_001369987.1 NP_001356921.1 2 - 3 607 68
NM_169927.3 NP_732557.2 2 - 3 607 68

View Full Product Details