Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- lqfR
- Entrez Gene ID
-
42632
- Gene Name
- liquid facets-Related
- Gene Aliases
-
CG13853, CG13854, CG31170, CG31285, CG42250, Dmel\CG42250, Dmel_CG42250, Epsin, LqfR, Lqfr, Tel2, XII-10, dTel2, epsin, epsin-like, epsin02, epsinR, l(3)03685, l(3)A9, l(3)SG62, l(3)XII-10, l(3)dsl-9, l(3)dsl9
- Chromosome Location
-
Chr.3R: 22411301 - 22419051 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261279
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170009.4 |
NP_732734.3 |
5 - 6 |
1762 |
64 |
GenBank mRNA |
AY060606.1 |
- |
5 - 6 |
1730 |
64 |
View Full Product Details