Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- btn
- Entrez Gene ID
-
42664
- Gene Name
- buttonless
- Gene Aliases
-
1261/13, BTN, CG5264, Dmel\CG5264, Dmel_CG5264, btn/MEOX
- Chromosome Location
-
Chr.3R: 22586149 - 22586625 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0014949
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170026.2 |
NP_732768.1 |
1 - 1 |
173 |
89 |
View Full Product Details