Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnc
Entrez Gene ID
42743
Gene Name
cap-n-collar
Gene Aliases
5134, BcDNA:RE05559, CG13826, CG17894, CG43286, CG4566, CG4578, CNC, CNC_DROME, CnC, Cnc, Cnc-C, CncC, DM12, Dmel\CG43286, Dmel_CG43286, NRF2, Nrf2, anon-WO0153538.6, anon-WO0153538.7, cnc-C, cncC, cncC/Nrf2, dNrf2, l(3)03921, l(3)j5E7, nrf2
Chromosome Location
Chr.3R: 23185580 - 23226711 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262975
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260327.2 NP_001247256.1 2 - 3 1355 65
NM_001260328.1 NP_001247257.1 2 - 3 1332 65
NM_001260329.2 NP_001247258.1 9 - 10 3362 65
NM_001260330.1 NP_001247259.1 10 - 11 4565 65
NM_001260331.1 NP_001247260.1 10 - 11 4734 65
NM_001260332.2 NP_001247261.1 9 - 10 3349 65
NM_001260333.2 NP_001247262.1 9 - 10 3358 65
NM_001275935.1 NP_001262864.1 10 - 11 4481 65
NM_001275936.1 NP_001262865.1 4 - 5 2041 65
NM_170053.3 NP_732833.1 10 - 11 4481 65
NM_170054.4 NP_732834.1 6 - 7 2960 65
NM_170055.2 NP_732835.1 1 - 2 1257 65
NM_170056.2 NP_732836.1 2 - 3 1350 65
NM_170057.2 NP_732837.1 2 - 3 1330 65
NM_170058.2 NP_732838.2 8 - 9 2899 65
NM_170059.3 NP_732839.1 2 - 3 1701 65
GenBank mRNA AY061154.1 - 2 - 3 1355 65

View Full Product Details