Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sba
Entrez Gene ID
42824
Gene Name
six-banded
Gene Aliases
AAF56201, CG13598, DMTF125, Dm Sba, Dmel\CG13598, Dmel_CG13598, F125, TF125, Tf125
Chromosome Location
Chr.3R: 23894174 - 23922973 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0016754
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104422.3 NP_001097892.1 6 - 7 4020 67
NM_001104423.2 NP_001097893.1 6 - 7 3949 67
NM_001260342.2 NP_001247271.1 6 - 7 3949 67
NM_001275959.1 NP_001262888.1 6 - 7 3949 67
NM_001275960.1 NP_001262889.1 6 - 7 4020 67
NM_170094.4 NP_732911.2 5 - 6 3757 67
NM_176549.3 NP_788726.2 5 - 6 3757 67

View Full Product Details