Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cav
Entrez Gene ID
42881
Gene Name
caravaggio
Gene Aliases
1G5, Anon1G5, CG6219, Caravaggio, Cav, Dmel\CG6219, Dmel_CG6219, HOAP, Hoap, anon fe 1G5, anon-EST:fe1G5, anon1G5, cav/HOAP, hoap, p55
Chromosome Location
Chr.3R: 24221103 - 24223528 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0026257
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079753.4 NP_524477.3 2 - 3 529 93
NM_170133.2 NP_732969.1 1 - 2 235 93
NM_170134.2 NP_732970.1 2 - 3 658 93
NM_170135.2 NP_732971.2 2 - 3 785 93
GenBank mRNA AY061114.1 - 2 - 3 535 93
BT125833.1 - 2 - 3 277 93

View Full Product Details