Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
jar
Entrez Gene ID
42889
Gene Name
jaguar
Gene Aliases
95F, 95F MHC, CG5695, Dm 95F, Dm95F, Dmel\CG5695, Dmel_CG5695, Dro95F, JAG, Jaguar, Jar, M6, MHC95F, MYOVI, Mhc95F, Myo1A, MyoVI, Myosin VI, dMyoVI, jag, ms(3)06746, myo VI, myoVI, myosins VI
Chromosome Location
Chr.3R: 24252826 - 24271233 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011225
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014647.3 NP_001014647.2 11 - 12 3209 58
NM_001014648.3 NP_001014648.2 11 - 12 3209 58
NM_001014649.3 NP_001014649.2 11 - 12 3209 58
NM_001014650.3 NP_001014650.2 11 - 12 3209 58
NM_001275976.1 NP_001262905.1 11 - 12 2967 58
NM_001275977.2 NP_001262906.1 11 - 12 3676 58
NM_001300572.1 NP_001287501.1 11 - 12 3209 58
NM_079754.4 NP_524478.4 11 - 12 3209 58
NM_170138.3 NP_732976.1 11 - 12 2967 58
GenBank mRNA BT133083.1 - 11 - 12 3206 58

View Full Product Details