Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ash2
Entrez Gene ID
42936
Gene Name
absent, small, or homeotic discs 2
Gene Aliases
1124/11, 291.8, 703, ASH2, Ash2, CG6677, Dmel\CG6677, Dmel_CG6677, ash-2, l(3)112411, l(3)S112411, l(3)SG65, mad
Chromosome Location
Chr.3R: 24650125 - 24653376 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000139
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176558.3 NP_788735.1 1 - 2 223 87
GenBank mRNA AY051803.1 - 1 - 2 176 87
U73809.2 - 1 - 2 176 87

View Full Product Details