Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
msi
Entrez Gene ID
43087
Gene Name
musashi
Gene Aliases
CG5099, DMSIDNA, Dmel\CG5099, Dmel_CG5099, MSI, Msi, SIDNA, anon-EST:Liang-2.35, clone 2.35, dMSI, dMsi
Chromosome Location
Chr.3R: 25516118 - 25608423 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011666
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260374.2 NP_001247303.1 3 - 4 743 70
NM_001276031.1 NP_001262960.1 3 - 4 743 70
NM_170229.3 NP_733108.2 3 - 4 661 70
GenBank mRNA BT133366.1 - 3 - 4 661 70

View Full Product Details