Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
msi
Entrez Gene ID
43087
Gene Name
musashi
Gene Aliases
CG5099, DMSIDNA, Dmel\CG5099, Dmel_CG5099, MSI, Msi, SIDNA, anon-EST:Liang-2.35, clone 2.35, dMSI, dMsi
Chromosome Location
Chr.3R: 25516118 - 25608423 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011666
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260374.2 NP_001247303.1 6 - 7 2016 109
NM_001260375.2 NP_001247304.1 4 - 5 1929 109
NM_001276030.1 NP_001262959.1 5 - 6 1968 109
NM_001276031.1 NP_001262960.1 6 - 7 2016 109
NM_001276032.1 NP_001262961.1 4 - 5 1804 109
NM_079838.4 NP_524577.1 2 - 3 1827 109
NM_170229.3 NP_733108.2 6 - 7 1934 109
GenBank mRNA AY051802.1 - 2 - 3 1789 109
BT133366.1 - 6 - 7 1934 109

View Full Product Details