Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LpR1
Entrez Gene ID
2768687
Gene Name
Lipophorin receptor 1
Gene Aliases
CG31094, CG31095, CG4861, CG4870, CG4874, CT15569, Dmel\CG31094, Dmel_CG31094, lpr1
Chromosome Location
Chr.3R: 25741348 - 25763105 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0066101
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104461.3 NP_001097931.3 8 - 9 2581 131
NM_001104463.3 NP_001097933.2 8 - 9 2893 131
NM_001104464.2 NP_001097934.2 10 - 11 3133 131
NM_170240.3 NP_733119.2 8 - 9 2587 131
NM_206573.2 NP_996296.2 9 - 10 3016 131
GenBank mRNA BT011057.1 - 8 - 9 2893 131
BT011063.1 - 9 - 10 2995 131

View Full Product Details