Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Hex-t2
- Entrez Gene ID
-
43191
- Gene Name
- CG32849-PA
- Gene Aliases
-
CG32849, CG5443, Dmel\CG32849, Dmel_CG32849, HK, Hex, Hex-T2, anon-WO0140519.203
- Chromosome Location
-
Chr.3R: 26450671 - 26452327 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0042710
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170272.3 |
NP_733151.2 |
1 - 2 |
623 |
57 |
View Full Product Details