Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nAChRalpha1
Entrez Gene ID
42918
Gene Name
nicotinic Acetylcholine Receptor alpha1
Gene Aliases
AChRalpha1, ALS, ALs, Acr96A, Acr96Aa, AcrB, CG5610, CG5610-PA, Dalpha1, Dalpha1 nAChR, Dmel\CG5610, Dmel_CG5610, EMS1, NAcRa96Aa, alpha-like, alpha1, als, dALS, nAChR, nAChR-alpha96Aa, nAChRalpha-96Aa, nAcRalpha-96Aa, nAcRs, nAcr96Aa
Chromosome Location
Chr.3R: 24393896 - 24457157 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000036
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275987.1 NP_001262916.1 5 - 6 1830 85
NM_001275988.2 NP_001262917.1 5 - 6 1830 85
NM_079757.4 NP_524481.2 5 - 6 1830 85

View Full Product Details