Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- asp
- Entrez Gene ID
-
42946
- Gene Name
- abnormal spindle
- Gene Aliases
-
ASP, Asp, CG6875, Dm Asp, Dmel\CG6875, Dmel_CG6875, anon-96Aa, anon-WO0118547.279
- Chromosome Location
-
Chr.3R: 24753849 - 24760513 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000140
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079764.3 |
NP_524488.3 |
2 - 3 |
379 |
64 |
GenBank mRNA |
BT132945.1 |
- |
2 - 3 |
325 |
64 |
View Full Product Details