Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Stt3B
- Entrez Gene ID
-
43005
- Gene Name
- Catalytic subunit 3B of the oligosaccharyltransferase complex
- Gene Aliases
-
BcDNA.GM01838, BcDNA:GM01838, CG7748, Dmel\CG7748, Dmel_CG7748, OstStt3, STT, anon-WO03054008.7, l(3)j2D9
- Chromosome Location
-
Chr.3R: 25066712 - 25069637 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0011336
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079770.3 |
NP_524494.1 |
3 - 4 |
2136 |
99 |
View Full Product Details