Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- E(spl)mgamma-HLH
- Entrez Gene ID
-
43151
- Gene Name
- Enhancer of split mgamma, helix-loop-helix
- Gene Aliases
-
CG8333, Dm-mB, Dmel\CG8333, Dmel_CG8333, E(Spl) HLH-m gamma, E(spl) m gamma, E(spl) mg, E(spl) mgamma, E(spl)-mgamma, E(spl)Mgamma, E(spl)gamma, E(spl)m-gamma, E(spl)mB, E(spl)mgamma, HLH-mgamma, HLHMgamma, HLHmB, HLHmgamma, HlHmg, Mgamma, anon-EST:Liang-1.49, bHLHb34, clone 1.49, e(spl)mgamma, gamma, mB, mg, mgamma
- Chromosome Location
-
Chr.3R: 25999851 - 26000761 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002735
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079780.3 |
NP_524504.2 |
1 - 1 |
785 |
114 |
GenBank mRNA |
BT022125.2 |
- |
1 - 1 |
754 |
114 |
View Full Product Details