Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RYa-R
- Entrez Gene ID
-
43253
- Gene Name
- RYamide receptor
- Gene Aliases
-
ATR, CG5811, CG5811/107368, Dm RYa-R, Dmel\CG5811, Dmel_CG5811, NepYR, NepYr, PR4, anon-WO0131005.13, anon-WO0131005.15, dNepYr
- Chromosome Location
-
Chr.3R: 26996980 - 27057935 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004842
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079801.4 |
NP_524525.3 |
6 - 7 |
1729 |
73 |
View Full Product Details