Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
spz
Entrez Gene ID
43256
Gene Name
spatzle
Gene Aliases
CG6134, CT19282, Dmel\CG6134, Dmel_CG6134, SPZ, Spz, Spz-1, Spz1, mel(3)7
Chromosome Location
Chr.3R: 27064990 - 27070070 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003495
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104472.2 NP_001097942.1 5 - 6 1558 57
NM_001144644.2 NP_001138116.1 4 - 5 1498 57
NM_079802.3 NP_524526.1 4 - 5 1666 57
NM_170309.2 NP_733188.1 3 - 4 1369 57
NM_170312.2 NP_733191.1 3 - 4 1588 57
NM_170313.2 NP_733192.1 4 - 5 1420 57
NM_170315.2 NP_733194.1 4 - 5 1447 57
NM_170316.2 NP_733195.1 4 - 5 1507 57
GenBank mRNA AF237968.1 - 4 - 5 586 57
AF237969.1 - 4 - 5 805 57
EF155533.1 - 4 - 5 637 57

View Full Product Details