Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Gr98c
- Entrez Gene ID
-
117336
- Gene Name
- Gustatory receptor 98c
- Gene Aliases
-
98B.3, CG12884, CG31060, Dmel\CG31060, Dmel_CG31060, GR98B.3, GR98c, Gr98A1, Gr98A2, Gr98B3
- Chromosome Location
-
Chr.3R: 27544443 - 27545836 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0046886
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079807.3 |
NP_524531.2 |
2 - 3 |
947 |
96 |
View Full Product Details