Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klp98A
Entrez Gene ID
43310
Gene Name
Kinesin-like protein at 98A
Gene Aliases
CG5658, DmKlp98A, DmSNX23, Dmel\CG5658, Dmel_CG5658, KIF 16A, KIF16A/B, KLP6, KLP98A, Klp6, Klp98a
Chromosome Location
Chr.3R: 27547775 - 27559145 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004387
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260410.2 NP_001247339.1 6 - 7 2697 100
NM_079808.4 NP_524532.2 6 - 7 2697 100
GenBank mRNA BT132947.1 - 6 - 7 2666 100

View Full Product Details