Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Dop1R2
- Entrez Gene ID
-
43484
- Gene Name
- Dopamine 1-like receptor 2
- Gene Aliases
-
CG18741, CG7569, D2R, DAMB, DDR2, DOPR2, DOPR99B, DR2, Damb, Dar2, DmDOP2, Dmel\CG18741, Dmel_CG18741, DopD1, DopR, DopR2, DopR99B, DopRII, K13sc, damb, dopr2
- Chromosome Location
-
Chr.3R: 29630304 - 29659984 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266137
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079824.4 |
NP_524548.1 |
4 - 5 |
2434 |
71 |
GenBank mRNA |
U34383.1 |
- |
2 - 3 |
|
71 |
View Full Product Details