Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ocn
Entrez Gene ID
43567
Gene Name
ocnus
Gene Aliases
BcDNA:GH02250, CG7929, Dmel\CG7929, Dmel_CG7929, Ocn, bs29e10.y1
Chromosome Location
Chr.3R: 30037003 - 30037730 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041102
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079839.3 NP_524578.1 2 - 3 424 107
GenBank mRNA AF145597.1 - 2 - 3 412 107
KX532136.1 - 2 - 3 339 107

View Full Product Details