Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sap-r
Entrez Gene ID
43662
Gene Name
Saposin-related
Gene Aliases
Apa, BcDNA:GH08312, CG12070, DAPA, Dmel\CG12070, Dmel_CG12070, Ov9, P110, Sap-R, Sapr, anon-EST:ParkEST270, sap-r
Chromosome Location
Chr.3R: 30883464 - 30889062 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000416
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079858.4 NP_524597.1 3 - 4 539 76
NM_170529.2 NP_733408.2 2 - 3 448 76
GenBank mRNA AF145647.1 - 3 - 4 527 76
DQ062780.1 - 3 - 4 410 76

View Full Product Details