Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CycG
Entrez Gene ID
43724
Gene Name
Cyclin G
Gene Aliases
CG 11525, CG11525, Dmel\CG11525, Dmel_CG11525, anon-EST:Liang-1.25, anon-EST:Liang-2.24, chr3R:27424276..27424475, clone 1.25, clone 2.24, cycG, dCycG, i205, i261
Chromosome Location
Chr.3R: 31596642 - 31612664 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039858
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276217.2 NP_001263146.1 5 - 6 1746 117
NM_079870.4 NP_524609.2 5 - 6 1746 117
NM_170555.3 NP_733434.1 5 - 6 2589 117
NM_170556.3 NP_733435.1 5 - 6 2144 117
NM_170557.3 NP_733436.1 5 - 6 2002 117
GenBank mRNA AY058720.1 - 5 - 6 2595 117

View Full Product Details