Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
faf
Entrez Gene ID
43749
Gene Name
fat facets
Gene Aliases
BcDNA.LD22582, BcDNA:LD22582, CG1945, Dmel\CG1945, Dmel_CG1945, Faf, Fat, P55824, USPX
Chromosome Location
Chr.3R: 31777020 - 31790681 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0005632
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260459.1 NP_001247388.1 16 - 17 8208 67
NM_001260460.2 NP_001247389.2 16 - 17 8176 67
NM_079873.5 NP_524612.2 16 - 17 8176 67
NM_170576.3 NP_733455.1 16 - 17 8176 67

View Full Product Details