Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cas
Entrez Gene ID
44018
Gene Name
castor
Gene Aliases
CAS, CG2102, Cas, Dmel\CG2102, Dmel_CG2102, l(3)j1C2, l(3)neo33, ming
Chromosome Location
Chr.3R: 5712975 - 5716805 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004878
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275371.1 NP_001262300.1 3 - 4 1437 67
NM_079938.4 NP_524677.1 2 - 3 1743 67
GenBank mRNA AY051865.1 - 2 - 3 1721 67

View Full Product Details