Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ND-23
- Entrez Gene ID
-
44207
- Gene Name
- NADH dehydrogenase (ubiquinone) 23 kDa subunit
- Gene Aliases
-
CG3944, Dmel\CG3944, Dmel_CG3944, ND 23, ND23, NUIM
- Chromosome Location
-
Chr.3R: 15836048 - 15837117 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0017567
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079980.4 |
NP_524719.1 |
2 - 3 |
581 |
77 |
GenBank mRNA |
AY060840.1 |
- |
2 - 3 |
535 |
77 |
View Full Product Details