Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
disp
Entrez Gene ID
44274
Gene Name
dispatched
Gene Aliases
CG2019, Disp, Dmel\CG2019, Dmel_CG2019, anon-WO0146227.1
Chromosome Location
Chr.3R: 5849697 - 5856311 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0029088
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079995.3 NP_524734.2 4 - 5 2198 73
GenBank mRNA BT050429.1 - 4 - 5 2198 73

View Full Product Details