Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- cwo
- Entrez Gene ID
-
44669
- Gene Name
- clockwork orange
- Gene Aliases
-
AAF24476.1, CG17100, CWO, Cwo, Dmel\CG17100, Dmel_CG17100, GM05287, bHLHe49, dcwo, mel_cwo
- Chromosome Location
-
Chr.3R: 10388258 - 10400752 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0259938
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080036.3 |
NP_524775.1 |
1 - 2 |
1181 |
93 |
View Full Product Details