Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cwo
Entrez Gene ID
44669
Gene Name
clockwork orange
Gene Aliases
AAF24476.1, CG17100, CWO, Cwo, Dmel\CG17100, Dmel_CG17100, GM05287, bHLHe49, dcwo, mel_cwo
Chromosome Location
Chr.3R: 10388258 - 10400752 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0259938
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260096.2 NP_001247025.1 2 - 3 1764 70
NM_080036.3 NP_524775.1 2 - 3 1304 70
GenBank mRNA AY119588.1 - 1 - 2 118 70

View Full Product Details