Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fur1
Entrez Gene ID
47220
Gene Name
Furin 1
Gene Aliases
CG10772, Dfur1, Dfurin1, Dmel\CG10772, Dmel_CG10772, Furin, Furl, Klip1, dFur1, dFurin 1, dKLIP-1, fur-1, fur1, fur[1], furin, l(3)rL205
Chromosome Location
Chr.3R: 25347431 - 25473058 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004509
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276024.1 NP_001262953.1 8 - 9 2945 71
NM_001276025.1 NP_001262954.1 8 - 9 2945 71
NM_080146.3 NP_524885.1 8 - 9 2945 71
NM_170654.2 NP_733102.1 8 - 9 2945 71
NM_170655.2 NP_733103.1 8 - 9 2945 71
NM_170656.2 NP_733104.1 7 - 8 2880 71
NM_206570.2 NP_996293.1 9 - 10 3001 71
NM_206571.2 NP_996294.2 8 - 9 2945 71
GenBank mRNA AY069590.1 - 8 - 9 2927 71
L12372.1 - 8 - 9 2854 71
L12375.1 - 8 - 9 2920 71
L12376.1 - 7 - 8 2834 71

View Full Product Details