The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001275347.1 | NP_001262276.1 | 4 - 5 | 1196 | 69 |
NM_080258.4 | NP_524997.2 | 4 - 5 | 1097 | 69 | |
GenBank mRNA | BT133173.1 | - | 4 - 5 | 1106 | 69 |