Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- cpx
- Entrez Gene ID
-
64877
- Gene Name
- complexin
- Gene Aliases
-
BcDNA:GH27718, BcDNA:HL04936, BcDNA:RH26459, BcDNA:RH31085, CG32490, CPX, Cpx, DmCpx, Dmel\CG32490, Dmel_CG32490, dmCplx
- Chromosome Location
-
Chr.3R: 4280221 - 4304604 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0041605
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080280.4 |
NP_525019.2 |
5 - 6 |
806 |
78 |
View Full Product Details