Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- E(spl)m7-HLH
- Entrez Gene ID
-
43160
- Gene Name
- Enhancer of split m7, helix-loop-helix
- Gene Aliases
-
CG8361, Dmel\CG8361, Dmel_CG8361, E(SPL)m7, E(spl) m7, E(spl)-M7, E(spl)-m7, E(spl)M7, E(spl)m7, ESPL, En(spl)m7, HLH-m7, HLHm7, HlHm7, M7, bHLHb35, m7
- Chromosome Location
-
Chr.3R: 26037038 - 26038019 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002633
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_080505.3 |
NP_536753.1 |
1 - 1 |
674 |
93 |
GenBank mRNA |
BT022121.1 |
- |
1 - 1 |
716 |
93 |
View Full Product Details