Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ms
Entrez Gene ID
44324
Gene Name
Myosuppressin
Gene Aliases
CG6440, DMS, Dm-MS, Dmel\CG6440, Dmel_CG6440, Dms, Drm-MS, Drome-MS, FLRFa, MEMS, MS, Nems, cg6440, dms
Chromosome Location
Chr.3R: 24341551 - 24343825 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011581
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001275982.1 NP_001262911.1 2 - 3 325 85
NM_001300574.1 NP_001287503.1 2 - 3 342 85
NM_080511.3 NP_536772.1 2 - 3 342 85
GenBank mRNA AY075325.1 - 2 - 3 321 85
KX531969.1 - 1 - 2 211 85

View Full Product Details