Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppk31
Entrez Gene ID
318575
Gene Name
pickpocket 31
Gene Aliases
CG31065, Dmel\CG31065, Dmel_CG31065
Chromosome Location
Chr.3R: 27163124 - 27165131 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051065
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104474.1 NP_001097944.1 4 - 5 987 69

View Full Product Details