Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- tau
- Entrez Gene ID
-
326116
- Gene Name
- CG45110-PA
- Gene Aliases
-
BcDNA:RE16764, CG12881, CG31057, CG31058, CG34294, CG45110, CG5606, Dmel\CG45110, Dmel_CG45110, Tau, d-tau, dTau, dtau
- Chromosome Location
-
Chr.3R: 27639886 - 27656996 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266579
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170345.3 |
NP_733224.2 |
3 - 3 |
1672 |
74 |
View Full Product Details