Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jon99Ciii
Entrez Gene ID
43543
Gene Name
Jonah 99Ciii
Gene Aliases
99Calpha, CG15519, CG31362, Dmel\CG31362, Dmel_CG31362, Jon99B, Jon99C, Jon99C1, Jon99C2, Jon99Calpha, Jon99Calphaii, Jon99Cbeta, Jon99Cbetaiii, JonC, SER2, SP143, Ser2, Ser5, Ser99Db
Chromosome Location
Chr.3R: 29922216 - 29923455 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003357
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001300669.1 NP_001287598.1 1 - 1 846 76

View Full Product Details