Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PH4alphaEFB
- Entrez Gene ID
-
43620
- Gene Name
- prolyl-4-hydroxylase-alpha EFB
- Gene Aliases
-
BcDNA:SD05564, CG12088, CG1473, CG31022, Dmel\CG31022, Dmel_CG31022, PH4-aEFB, PH4alpha-EFB
- Chromosome Location
-
Chr.3R: 30466050 - 30485799 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0039776
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170492.2 |
NP_733371.1 |
7 - 8 |
1739 |
85 |
View Full Product Details