Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PH4alphaSG1
Entrez Gene ID
326112
Gene Name
prolyl-4-hydroxylase-alpha SG1
Gene Aliases
CG15542, CG31014, Dmel\CG31014, Dmel_CG31014, PHalphaSG1, Ph4alphaSG1, SG1, alphaSG1
Chromosome Location
Chr.3R: 30503665 - 30505661 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051014
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170497.4 NP_733376.1 1 - 2 196 112
GenBank mRNA BT023117.1 - 1 - 2 169 112

View Full Product Details