Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PH4alphaSG1
- Entrez Gene ID
-
326112
- Gene Name
- prolyl-4-hydroxylase-alpha SG1
- Gene Aliases
-
CG15542, CG31014, Dmel\CG31014, Dmel_CG31014, PHalphaSG1, Ph4alphaSG1, SG1, alphaSG1
- Chromosome Location
-
Chr.3R: 30503665 - 30505661 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0051014
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170497.4 |
NP_733376.1 |
2 - 3 |
879 |
73 |
GenBank mRNA |
BT023117.1 |
- |
2 - 3 |
852 |
73 |
View Full Product Details