Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- zfh1
- Entrez Gene ID
-
43650
- Gene Name
- Zn finger homeodomain 1
- Gene Aliases
-
CG1322, Dmel\CG1322, Dmel_CG1322, ZFH-1, ZFH1, ZFh1, Zfh-1, Zfh1, Zfh1a, l(3)00865, zfh-1, zfl-1, zhf1
- Chromosome Location
-
Chr.3R: 30765926 - 30789202 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004606
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170523.3 |
NP_733402.1 |
1 - 2 |
399 |
72 |
GenBank mRNA |
BT003277.1 |
- |
1 - 2 |
399 |
72 |
View Full Product Details