Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- pyd
- Entrez Gene ID
-
41062
- Gene Name
- polychaetoid
- Gene Aliases
-
CG11782, CG11962, CG12409, CG31349, CG43140, CG9729, CG9731, CG9763, CT36877, DZO-1, Dmel\CG43140, Dmel_CG43140, Pch, Pyd, Pyd/ZO-1, Tamou, ZO-1, ZO1, dzo-1, pyd(Z01), tam, xvt
- Chromosome Location
-
Chr.3R: 8827068 - 8931896 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0262614
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170646.6 |
NP_731293.3 |
1 - 2 |
1180 |
71 |
View Full Product Details