Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mical
Entrez Gene ID
41225
Gene Name
Molecule interacting with CasL
Gene Aliases
CG11685, CG11687, CG18484, CG18667, CG18668, CG33186, CG33190, CG33208, Dmel\CG33208, Dmel_CG33208, MICAL, mdcds_49226, mical, milch
Chromosome Location
Chr.3R: 10001554 - 10042895 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053208
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260085.2 NP_001247014.1 22 - 23 14372 66
NM_001260086.3 NP_001247015.1 23 - 24 14453 66
NM_001260087.3 NP_001247016.2 23 - 24 14693 66
NM_001316573.1 NP_001303502.1 21 - 22 14318 66
NM_001316574.1 NP_001303503.1 20 - 21 9224 66
NM_176443.2 NP_788620.1 19 - 20 8453 66
NM_176444.2 NP_788621.1 21 - 22 9257 66
NM_176445.2 NP_788622.1 20 - 21 14298 66
NM_176446.2 NP_788623.1 19 - 20 14254 66
NM_176447.2 NP_788624.1 23 - 24 14420 66
NM_176448.2 NP_788625.1 20 - 21 14194 66
NM_176449.3 NP_788626.2 19 - 20 14138 66

View Full Product Details