Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mical
Entrez Gene ID
41225
Gene Name
Molecule interacting with CasL
Gene Aliases
CG11685, CG11687, CG18484, CG18667, CG18668, CG33186, CG33190, CG33208, Dmel\CG33208, Dmel_CG33208, MICAL, mdcds_49226, mical, milch
Chromosome Location
Chr.3R: 10001554 - 10042895 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0053208
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144560.3 NP_001138032.1 3 - 4 1950 65
NM_001260085.2 NP_001247014.1 5 - 6 1997 65
NM_001260086.3 NP_001247015.1 5 - 6 2045 65
NM_001260087.3 NP_001247016.2 6 - 7 2072 65
NM_001316573.1 NP_001303502.1 6 - 7 2072 65
NM_001316574.1 NP_001303503.1 6 - 7 2072 65
NM_176443.2 NP_788620.1 6 - 7 2072 65
NM_176444.2 NP_788621.1 6 - 7 2072 65
NM_176445.2 NP_788622.1 3 - 4 1950 65
NM_176446.2 NP_788623.1 2 - 3 1906 65
NM_176447.2 NP_788624.1 6 - 7 2072 65
NM_176448.2 NP_788625.1 3 - 4 1846 65
NM_176449.3 NP_788626.2 3 - 4 1859 65
NM_176450.1 NP_788627.1 6 - 7 1998 65
GenBank mRNA AY118290.1 - 6 - 7 1998 65

View Full Product Details