Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- B52
- Entrez Gene ID
-
41670
- Gene Name
- B52-PA
- Gene Aliases
-
B52/SRp55, B52/dSRp55, CG10851, Dmel\CG10851, Dmel_CG10851, E(Dfd), E726, MabB52, RRM8, RSp55, Rbp8, SR55, SRSF6, SRp55, dSRp55, l(3)s2249, rrm8
- Chromosome Location
-
Chr.3R: 13661311 - 13666891 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004587
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176491.3 |
NP_788668.1 |
8 - 9 |
1110 |
102 |
View Full Product Details