Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mask
Entrez Gene ID
50070
Gene Name
multiple ankyrin repeats single KH domain
Gene Aliases
CG18671, CG31138, CG33106, CG6268, CG6313, Dmel\CG33106, Dmel_CG33106, EC3-6, MASK, Mask, cg6313
Chromosome Location
Chr.3R: 24230562 - 24250248 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0043884
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001260351.1 NP_001247280.1 16 - 17 11976 195
NM_001275974.1 NP_001262903.1 15 - 16 10782 195
NM_001275975.1 NP_001262904.1 15 - 16 11946 195
NM_001300571.1 NP_001287500.1 14 - 15 10743 195
NM_176556.2 NP_788733.1 15 - 16 11949 195
NM_176557.2 NP_788734.1 15 - 16 11949 195

View Full Product Details