Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
janA
Entrez Gene ID
43569
Gene Name
janus A
Gene Aliases
BcDNA:AT12574, CG7933, Dmel\CG7933, Dmel_CG7933, Jan, Y, jan, jan A, janus
Chromosome Location
Chr.3R: 30038622 - 30039533 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001280
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_176585.2 NP_788762.1 1 - 2 224 59
NM_176586.4 NP_788763.1 2 - 3 206 59
NM_206588.2 NP_996311.1 2 - 3 227 59

View Full Product Details