Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Atpalpha
- Entrez Gene ID
-
48971
- Gene Name
- Na pump alpha subunit
- Gene Aliases
-
(Na[+]/k[+])-ATPase, ATP-alpha, ATPBagr, ATPa, ATPalpha, Atp, Atp a, Atpa, CG5670, Da-47, Dmel\CG5670, Dmel_CG5670, JY, K ATPase, K-ATPase, K[+] -ATPase, K[+]-ATPase, K[+]-ATPase alpha subunit, K[+]-ATPase alpha-subunit, NKA, Na, Na+-K+-ATPase, Na+/K+ ATPase alpha subunit, Na+/K+-ATPase, Na-p, Na/K ATPase, Na/K-ATPase, Na/K-ATPase alpha-subunit, NaK, NaK ATPase, NaK-ATPase, Na[+], Na[+]-K[+]-ATPase alpha[[5]]-subunit, Na[+]/K[+] ATPase, Na[+]/K[+]-ATPase, Na[+]/K[+]ATPase, Na[+]K[+] ATPase, Na[+]K[+]-ATPase, Na[+]K[+]ATPase, alpha-ATPase, atpalpha, dnATPase, l(3)01164, l(3)01453, l(3)04492, l(3)j3B2, l(3)j3B7, l(3)j6A4, l(3)rM454, l(3)ry136, sodium/potassium transporting ATPase alpha
- Chromosome Location
-
Chr.3R: 20948714 - 20976239 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002921
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_206526.3 |
NP_996248.1 |
7 - 8 |
2643 |
76 |
View Full Product Details