Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCAP-R
Entrez Gene ID
2768688
Gene Name
Crustacean cardioactive peptide receptor
Gene Aliases
CCAP receptor, CCAPR, CG14547, CG33344, CG6111, CcapR, DCCAPR, Dmel\CG33344, Dmel_CG33344, anon-WO0170980.46, anon-WO0170980.47
Chromosome Location
Chr.3R: 25923499 - 25956270 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039396
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382065.1 NP_001368957.1 2 - 3 1009 74
NM_206574.4 NP_996297.3 2 - 3 1009 74
GenBank mRNA BT133202.1 - 2 - 3 1009 74

View Full Product Details